The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182519.2 | NP_872325.2 | 10 - 11 | 1410 | 91 |
GenBank mRNA | AF549190.1 | - | 9 - 10 | 1278 | 91 |