Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD1B
Entrez Gene ID
58490
Gene Name
regulation of nuclear pre-mRNA domain containing 1B
Gene Aliases
C20orf77, CREPT, NET60, dJ1057B20.2
Chromosome Location
Chr.20: 38033462 - 38092366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021215.3 NP_067038.1 5 - 6 1058 115
XM_005260477.3 XP_005260534.1 5 - 6 1142 115
XM_005260480.4 XP_005260537.1 5 - 6 1279 115
XM_011528954.2 XP_011527256.1 5 - 6 1142 115
XM_017027990.1 XP_016883479.1 5 - 6 1142 115
XM_017027991.1 XP_016883480.1 4 - 5 1008 115
GenBank mRNA AK312468.1 - 5 - 6 818 115
BC033629.1 - 5 - 6 842 115
DQ372938.1 - 5 - 6 931 115

View Full Product Details