The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022900.4 | NP_075051.4 | 2 - 3 | 455 | 116 |
XM_006716093.3 | XP_006716156.2 | 2 - 3 | 293 | 116 | |
XM_011516495.2 | XP_011514797.1 | 2 - 3 | 293 | 116 | |
XM_017012546.1 | XP_016868035.1 | 2 - 3 | 293 | 116 | |
XM_017012547.1 | XP_016868036.1 | 2 - 3 | 255 | 116 | |
GenBank mRNA | AF355594.1 | - | 2 - 3 | 248 | 116 |
AF397424.2 | - | 2 - 3 | 375 | 116 | |
AK123866.1 | - | 2 - 3 | 293 | 116 | |
AK314329.1 | - | 2 - 3 | 253 | 116 | |
BC063284.1 | - | 2 - 3 | 440 | 116 |