Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNMBP
Entrez Gene ID
23268
Gene Name
dynamin binding protein
Gene Aliases
ARHGEF36, TUBA
Chromosome Location
Chr.10: 99875571 - 100009953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318326.1 NP_001305255.1 11 - 12 2927 63
NM_001318327.1 NP_001305256.1 11 - 12 1678 63
NM_015221.3 NP_056036.1 14 - 15 3924 63
XM_006717735.2 XP_006717798.1 13 - 14 3790 63
XM_011539559.2 XP_011537861.1 15 - 16 3995 63
GenBank mRNA AB023227.1 - 11 - 12 3014 63
AK128576.1 - 11 - 12 2926 63
AK303479.1 - 6 - 7 1025 63
AL833283.1 - 14 - 15 3890 63
AY196211.1 - 13 - 14 3798 63
BC041628.1 - 11 - 12 1667 63

View Full Product Details