Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNMBP
Entrez Gene ID
23268
Gene Name
dynamin binding protein
Gene Aliases
ARHGEF36, TUBA
Chromosome Location
Chr.10: 99875571 - 100009953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318326.1 NP_001305255.1 9 - 10 2420 95
NM_001318327.1 NP_001305256.1 9 - 10 1171 95
NM_015221.3 NP_056036.1 12 - 13 3417 95
XM_011539559.2 XP_011537861.1 13 - 14 3488 95
GenBank mRNA AB023227.1 - 9 - 10 2507 95
AK128576.1 - 9 - 10 2419 95
AK303479.1 - 4 - 5 518 95
AY196211.1 - 11 - 12 3291 95
BC041628.1 - 9 - 10 1160 95

View Full Product Details