Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCF2
Entrez Gene ID
4688
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
NCF-2, NOXA2, P67-PHOX, P67PHOX
Chromosome Location
Chr.1: 183555562 - 183590921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000433.3 NP_000424.2 4 - 5 775 72
NM_001127651.2 NP_001121123.1 5 - 6 612 72
XM_011509580.1 XP_011507882.1 5 - 6 613 72
XM_011509581.1 XP_011507883.1 5 - 6 600 72
GenBank mRNA AB209647.1 - 4 - 5 655 72
AF527950.1 - 4 - 5 522 72
AK312666.1 - 4 - 5 685 72
BC001606.1 - 4 - 5 752 72
BP193903.1 - 5 - 6 459 72
BT007439.1 - 4 - 5 500 72
KU178216.1 - 4 - 5 500 72
M32011.1 - 5 - 6 567 72

View Full Product Details