The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166108.1 | NP_001159580.1 | 14 - 15 | 2682 | 116 |
NM_001166109.1 | NP_001159581.1 | 12 - 13 | 1796 | 116 | |
NM_001166110.1 | NP_001159582.1 | 5 - 6 | 1295 | 116 | |
NM_016081.3 | NP_057165.3 | 13 - 14 | 2631 | 116 | |
XM_005262861.4 | XP_005262918.1 | 15 - 16 | 3381 | 116 | |
XM_005262866.2 | XP_005262923.1 | 14 - 15 | 2931 | 116 | |
XM_011531768.2 | XP_011530070.1 | 16 - 17 | 3635 | 116 | |
XM_011531769.2 | XP_011530071.1 | 15 - 16 | 3584 | 116 | |
XM_011531770.2 | XP_011530072.1 | 16 - 17 | 3635 | 116 | |
XM_011531773.1 | XP_011530075.1 | 15 - 16 | 2963 | 116 | |
XM_011531774.1 | XP_011530076.1 | 14 - 15 | 2912 | 116 | |
XM_011531775.1 | XP_011530077.1 | 14 - 15 | 2316 | 116 | |
XM_017007910.1 | XP_016863399.1 | 15 - 16 | 3584 | 116 | |
GenBank mRNA | AB023209.1 | - | 5 - 6 | 1259 | 116 |
AF077041.1 | - | 1 - 2 | 745 | 116 | |
AF464873.1 | - | 13 - 14 | 2631 | 116 | |
AK000845.1 | - | 1 - 2 | 220 | 116 | |
AK025843.1 | - | 1 - 2 | 222 | 116 | |
AK095512.1 | - | 12 - 13 | 1758 | 116 | |
AY211921.1 | - | 4 - 5 | 481 | 116 | |
BC013867.2 | - | 4 - 5 | 475 | 116 | |
BC140850.1 | - | 13 - 14 | 2593 | 116 | |
BC144666.1 | - | 14 - 15 | 2644 | 116 |