The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166108.1 | NP_001159580.1 | 7 - 8 | 1681 | 80 |
NM_001166109.1 | NP_001159581.1 | 6 - 7 | 846 | 80 | |
NM_016081.3 | NP_057165.3 | 7 - 8 | 1681 | 80 | |
XM_005262861.4 | XP_005262918.1 | 7 - 8 | 1708 | 80 | |
XM_005262866.2 | XP_005262923.1 | 6 - 7 | 1258 | 80 | |
XM_011531768.2 | XP_011530070.1 | 8 - 9 | 1962 | 80 | |
XM_011531769.2 | XP_011530071.1 | 8 - 9 | 1962 | 80 | |
XM_011531770.2 | XP_011530072.1 | 8 - 9 | 1962 | 80 | |
XM_011531771.2 | XP_011530073.1 | 8 - 9 | 1962 | 80 | |
XM_011531772.2 | XP_011530074.1 | 8 - 9 | 1962 | 80 | |
XM_011531773.1 | XP_011530075.1 | 8 - 9 | 1962 | 80 | |
XM_011531774.1 | XP_011530076.1 | 8 - 9 | 1962 | 80 | |
XM_011531775.1 | XP_011530077.1 | 6 - 7 | 643 | 80 | |
XM_017007910.1 | XP_016863399.1 | 8 - 9 | 1962 | 80 | |
XM_017007911.1 | XP_016863400.1 | 8 - 9 | 1962 | 80 | |
GenBank mRNA | AF464873.1 | - | 7 - 8 | 1681 | 80 |
AK095512.1 | - | 6 - 7 | 808 | 80 | |
BC140850.1 | - | 7 - 8 | 1643 | 80 | |
BC144666.1 | - | 7 - 8 | 1643 | 80 |