The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024646.2 | NP_078922.1 | 9 - 10 | 1835 | 113 |
XM_006710898.3 | XP_006710961.1 | 9 - 10 | 1722 | 113 | |
XM_017002336.1 | XP_016857825.1 | 9 - 10 | 1840 | 113 | |
GenBank mRNA | AB051517.1 | - | 9 - 10 | 1785 | 113 |
AK023518.1 | - | 2 - 3 | 203 | 113 | |
AK056645.1 | - | 7 - 8 | 779 | 113 | |
AK095643.1 | - | 8 - 9 | 1475 | 113 | |
BC029832.1 | - | 7 - 8 | 849 | 113 | |
BX647643.1 | - | 7 - 8 | 1008 | 113 | |
DQ646393.1 | - | 9 - 10 | 1854 | 113 |