The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017995.2 | NP_001017995.1 | 5 - 6 | 578 | 82 |
NM_001308175.1 | NP_001295104.1 | 5 - 6 | 578 | 82 | |
XM_017009351.1 | XP_016864840.1 | 5 - 6 | 578 | 82 | |
GenBank mRNA | AB430862.1 | - | 5 - 6 | 407 | 82 |
BC038561.1 | - | 5 - 6 | 413 | 82 | |
DQ109556.1 | - | 5 - 6 | 413 | 82 |