The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 7 - 8 | 902 | 84 |
XM_005256045.3 | XP_005256102.1 | 6 - 7 | 787 | 84 | |
XM_011523227.2 | XP_011521529.1 | 7 - 8 | 1632 | 84 | |
GenBank mRNA | AF175265.1 | - | 7 - 8 | 860 | 84 |
AF183418.1 | - | 6 - 7 | 811 | 84 | |
AF186382.1 | - | 7 - 8 | 856 | 84 | |
AF191298.2 | - | 7 - 8 | 830 | 84 | |
AK001614.1 | - | 7 - 8 | 850 | 84 | |
AK023650.1 | - | 3 - 4 | 413 | 84 | |
AK025774.1 | - | - | 2510 | 84 | |
AK223218.1 | - | 7 - 8 | 821 | 84 | |
AL136888.1 | - | 7 - 8 | 850 | 84 | |
BC002414.2 | - | 6 - 7 | 809 | 84 | |
BC010362.1 | - | 7 - 8 | 823 | 84 | |
BC093036.1 | - | 7 - 8 | 848 | 84 | |
BX647213.1 | - | 2 - 3 | 1192 | 84 |