The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 8 - 9 | 1005 | 91 |
XM_005256045.3 | XP_005256102.1 | 7 - 8 | 890 | 91 | |
XM_011523227.2 | XP_011521529.1 | 8 - 9 | 1735 | 91 | |
GenBank mRNA | AF175265.1 | - | 8 - 9 | 963 | 91 |
AF183418.1 | - | 7 - 8 | 914 | 91 | |
AF186382.1 | - | 8 - 9 | 959 | 91 | |
AF191298.2 | - | 8 - 9 | 933 | 91 | |
AK001614.1 | - | 8 - 9 | 953 | 91 | |
AK023650.1 | - | 4 - 5 | 516 | 91 | |
AK025774.1 | - | - | 2613 | 91 | |
AK223218.1 | - | 8 - 9 | 924 | 91 | |
AL136888.1 | - | 8 - 9 | 953 | 91 | |
BC002414.2 | - | 7 - 8 | 912 | 91 | |
BC010362.1 | - | 8 - 9 | 926 | 91 | |
BC093036.1 | - | 8 - 9 | 951 | 91 | |
BX647213.1 | - | 3 - 4 | 1295 | 91 |