The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 9 - 10 | 1114 | 104 |
XM_005256045.3 | XP_005256102.1 | 8 - 9 | 999 | 104 | |
XM_011523227.2 | XP_011521529.1 | 9 - 10 | 1844 | 104 | |
GenBank mRNA | AF175265.1 | - | 9 - 10 | 1072 | 104 |
AF183418.1 | - | 8 - 9 | 1023 | 104 | |
AF186382.1 | - | 9 - 10 | 1068 | 104 | |
AF191298.2 | - | 9 - 10 | 1042 | 104 | |
AK001614.1 | - | 9 - 10 | 1062 | 104 | |
AK023650.1 | - | 5 - 6 | 625 | 104 | |
AK025774.1 | - | - | 2722 | 104 | |
AK223218.1 | - | 9 - 10 | 1033 | 104 | |
AL136888.1 | - | 9 - 10 | 1062 | 104 | |
AL512769.1 | - | 2 - 3 | 119 | 104 | |
BC002414.2 | - | 8 - 9 | 1021 | 104 | |
BC010362.1 | - | 9 - 10 | 1035 | 104 | |
BC093036.1 | - | 9 - 10 | 1060 | 104 |