Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LBP
Entrez Gene ID
3929
Gene Name
lipopolysaccharide binding protein
Gene Aliases
BPIFD2
Chromosome Location
Chr.20: 38346411 - 38377011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004139.4 NP_004130.2 4 - 5 629 88
GenBank mRNA AF105067.1 - 4 - 5 540 88
AK313625.1 - 4 - 5 559 88
AY344471.1 - 4 - 5 546 88
BC022256.1 - 4 - 5 558 88

View Full Product Details