The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004139.4 | NP_004130.2 | 6 - 7 | 757 | 103 |
GenBank mRNA | AF105067.1 | - | 6 - 7 | 668 | 103 |
AK313625.1 | - | 6 - 7 | 687 | 103 | |
BC022256.1 | - | 6 - 7 | 686 | 103 | |
M35533.1 | - | 6 - 7 | 651 | 103 |