The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006690.3 | NP_006681.1 | 3 - 4 | 510 | 75 |
XM_011528500.2 | XP_011526802.1 | 3 - 4 | 507 | 75 | |
XM_017027597.1 | XP_016883086.1 | 3 - 4 | 507 | 75 | |
XM_017027598.1 | XP_016883087.1 | 3 - 4 | 481 | 75 | |
GenBank mRNA | AB021227.1 | - | 3 - 4 | 510 | 75 |
AF131284.1 | - | 3 - 4 | 507 | 75 | |
BC047614.1 | - | 3 - 4 | 450 | 75 | |
DQ300264.1 | - | 4 - 5 | 351 | 75 |