The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006690.3 | NP_006681.1 | 4 - 5 | 820 | 58 |
XM_011528500.2 | XP_011526802.1 | 4 - 5 | 817 | 58 | |
XM_017027598.1 | XP_016883087.1 | 4 - 5 | 791 | 58 | |
GenBank mRNA | AF131284.1 | - | 4 - 5 | 817 | 58 |
BC047614.1 | - | 4 - 5 | 760 | 58 | |
DQ300264.1 | - | 5 - 6 | 661 | 58 |