The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022776.4 | NP_073613.2 | 3 - 4 | 1147 | 85 |
GenBank mRNA | AF346292.1 | - | 3 - 4 | 574 | 85 |
AF392454.1 | - | 3 - 4 | 718 | 85 | |
AK292702.1 | - | 3 - 4 | 499 | 85 | |
BC065213.1 | - | 3 - 4 | 497 | 85 |