Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
Chromosome Location
Chr.3: 125528858 - 125595537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022776.4 NP_073613.2 6 - 7 1609 99
GenBank mRNA AF346292.1 - 6 - 7 1036 99
AF392454.1 - 6 - 7 1180 99
AK023226.1 - 1 - 2 39 99
AK292702.1 - 6 - 7 961 99
BC010422.1 - 1 - 2 193 99
BC065213.1 - 6 - 7 959 99

View Full Product Details