The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287396.1 | NP_001274325.1 | 1 - 2 | 383 | 119 |
NM_001287397.1 | NP_001274326.1 | 1 - 2 | 383 | 119 | |
NM_178508.4 | NP_848603.2 | 1 - 2 | 383 | 119 | |
GenBank mRNA | BC047919.1 | - | 1 - 2 | 264 | 119 |
BU194178.1 | - | 1 - 2 | 264 | 119 |