The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002980.2 | NP_002971.2 | 1 - 2 | 299 | 125 |
XM_011511621.2 | XP_011509923.1 | 1 - 2 | 299 | 125 | |
XM_017004671.1 | XP_016860160.1 | 1 - 2 | 299 | 125 | |
GenBank mRNA | AK314638.1 | - | 1 - 2 | 302 | 125 |
BC035757.1 | - | 1 - 2 | 338 | 125 | |
U13989.1 | - | 1 - 2 | 123 | 125 | |
U20178.1 | - | 1 - 2 | 130 | 125 | |
U28281.1 | - | 1 - 2 | 181 | 125 |