The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006080.2 | NP_006071.1 | 4 - 5 | 768 | 91 |
XM_005250110.3 | XP_005250167.1 | 7 - 8 | 963 | 91 | |
XM_005250111.4 | XP_005250168.1 | 6 - 7 | 803 | 91 | |
XM_006715839.3 | XP_006715902.1 | 5 - 6 | 706 | 91 | |
XM_011515734.2 | XP_011514036.1 | 7 - 8 | 788 | 91 | |
XM_017011673.1 | XP_016867162.1 | 5 - 6 | 623 | 91 | |
GenBank mRNA | AK289954.1 | - | 4 - 5 | 861 | 91 |
BC111416.1 | - | 4 - 5 | 467 | 91 | |
DR002146.1 | - | 4 - 5 | 606 | 91 | |
L26081.1 | - | 4 - 5 | 468 | 91 |