The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002444.2 | NP_002435.1 | 12 - 13 | 1768 | 72 |
XM_005262269.2 | XP_005262326.1 | 12 - 13 | 1789 | 72 | |
XM_011530959.1 | XP_011529261.1 | 12 - 13 | 1684 | 72 | |
XM_017029545.1 | XP_016885034.1 | 14 - 15 | 1912 | 72 | |
XM_017029546.1 | XP_016885035.1 | 13 - 14 | 1808 | 72 | |
GenBank mRNA | AK297165.1 | - | 5 - 6 | 549 | 72 |
AK309986.1 | - | 2 - 3 | 301 | 72 | |
BC017293.2 | - | 12 - 13 | 1721 | 72 | |
EU794636.1 | - | 12 - 13 | 1721 | 72 | |
M69066.1 | - | 12 - 13 | 1670 | 72 |