Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSN
Entrez Gene ID
4478
Gene Name
moesin
Gene Aliases
HEL70
Chromosome Location
Chr.X: 65588382 - 65741931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002444.2 NP_002435.1 6 - 7 899 120
XM_005262269.2 XP_005262326.1 6 - 7 920 120
XM_011530959.1 XP_011529261.1 6 - 7 815 120
XM_017029545.1 XP_016885034.1 8 - 9 1043 120
XM_017029546.1 XP_016885035.1 7 - 8 939 120
GenBank mRNA BC001112.2 - 6 - 7 871 120
BC011827.2 - 6 - 7 871 120
BC017293.2 - 6 - 7 852 120
EU794636.1 - 6 - 7 852 120
M69066.1 - 6 - 7 801 120

View Full Product Details