Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
Chromosome Location
Chr.1: 27355179 - 27368037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297609.1 NP_001284538.1 10 - 11 1933 102
NM_004672.4 NP_004663.3 11 - 12 1957 102
XM_017002771.1 XP_016858260.1 9 - 10 1338 102
XM_017002772.1 XP_016858261.1 11 - 12 3128 102
GenBank mRNA AB167411.1 - 11 - 12 1688 102
AB208805.1 - 10 - 11 1402 102
AF100318.1 - 8 - 9 1102 102
BC015914.1 - 7 - 8 859 102
BC109032.2 - 2 - 3 236 102
BC129950.1 - 3 - 4 387 102
BC129951.1 - 3 - 4 387 102

View Full Product Details