The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173495.2 | NP_775766.2 | 1 - 2 | 360 | 59 |
XM_011545449.2 | XP_011543751.1 | 2 - 3 | 445 | 59 | |
GenBank mRNA | AK054858.1 | - | 1 - 2 | 250 | 59 |
AK298796.1 | - | 1 - 2 | 363 | 59 | |
BC062344.1 | - | 2 - 3 | 446 | 59 | |
BC121061.2 | - | 1 - 2 | 207 | 59 | |
DC351268.1 | - | 1 - 2 | 355 | 59 | |
DC365753.1 | - | 1 - 2 | 361 | 59 | |
KJ534921.1 | - | 1 - 2 | 352 | 59 |