The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012300.1 | NP_001012300.1 | 8 - 9 | 918 | 63 |
NM_001278341.1 | NP_001265270.1 | 7 - 8 | 828 | 63 | |
NM_006337.4 | NP_006328.2 | 9 - 10 | 1087 | 63 | |
XM_005268572.3 | XP_005268629.1 | 10 - 11 | 1187 | 63 | |
XM_011537760.2 | XP_011536062.1 | 8 - 9 | 4979 | 63 | |
XM_017018689.1 | XP_016874178.1 | 9 - 10 | 1423 | 63 | |
XM_017018690.1 | XP_016874179.1 | 10 - 11 | 1508 | 63 | |
GenBank mRNA | AF015308.1 | - | 8 - 9 | 995 | 63 |
AF068007.1 | - | 8 - 9 | 1627 | 63 | |
AY336730.1 | - | 8 - 9 | 918 | 63 | |
BC011794.2 | - | 9 - 10 | 1070 | 63 | |
BX538079.1 | - | 7 - 8 | 815 | 63 |