Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCRS1
Entrez Gene ID
10445
Gene Name
microspherule protein 1
Gene Aliases
ICP22BP, INO80Q, MCRS2, MSP58, P78
Chromosome Location
Chr.12: 49558294 - 49572157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012300.1 NP_001012300.1 9 - 10 948 78
NM_001278341.1 NP_001265270.1 8 - 9 858 78
NM_006337.4 NP_006328.2 10 - 11 1117 78
XM_005268572.3 XP_005268629.1 11 - 12 1217 78
XM_011537760.2 XP_011536062.1 9 - 10 5009 78
XM_017018689.1 XP_016874178.1 10 - 11 1453 78
XM_017018690.1 XP_016874179.1 11 - 12 1538 78
GenBank mRNA AF015308.1 - 9 - 10 1025 78
AF068007.1 - 9 - 10 1657 78
AY336730.1 - 9 - 10 948 78
BC011794.2 - 10 - 11 1100 78
BX538079.1 - 8 - 9 845 78

View Full Product Details