The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013342.3 | NP_037474.1 | 1 - 2 | 430 | 73 |
GenBank mRNA | AF052052.1 | - | 2 - 3 | 303 | 73 |
AK310627.1 | - | 1 - 2 | 412 | 73 | |
AK310647.1 | - | 1 - 2 | 401 | 73 | |
BC004281.1 | - | 1 - 2 | 62 | 73 | |
BC007776.2 | - | 1 - 2 | 48 | 73 | |
BI820344.1 | - | 1 - 2 | 430 | 73 |