Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
79042
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
LENG5, PCH2C, SEN34, SEN34L
Chromosome Location
Chr.19: 54189517 - 54194536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077446.3 NP_001070914.1 2 - 3 641 89
NM_001282332.1 NP_001269261.1 3 - 4 801 89
NM_001282333.1 NP_001269262.1 3 - 4 598 89
NM_024075.4 NP_076980.2 3 - 4 677 89
XM_005278290.4 XP_005278347.1 3 - 4 593 89
XM_011527294.2 XP_011525596.1 3 - 4 572 89
XM_011527295.1 XP_011525597.1 3 - 4 574 89
GenBank mRNA AK025929.1 - 3 - 4 596 89
AK054944.1 - 1 - 2 1627 89
AK225385.1 - 3 - 4 596 89
AK226013.1 - 3 - 4 599 89
AK300054.1 - 3 - 4 579 89
BC004530.2 - 3 - 4 801 89
BC020805.1 - 2 - 3 581 89
DR763016.1 - 3 - 4 573 89

View Full Product Details