The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318029.1 | NP_001304958.1 | 2 - 3 | 657 | 119 |
NM_176875.3 | NP_795344.1 | 3 - 4 | 909 | 119 | |
XM_005253210.1 | XP_005253267.1 | 3 - 4 | 972 | 119 | |
XM_017018516.1 | XP_016874005.1 | 3 - 4 | 1395 | 119 | |
GenBank mRNA | AF239668.1 | - | 3 - 4 | 657 | 119 |
AF441129.1 | - | 3 - 4 | 654 | 119 | |
AK226020.1 | - | 3 - 4 | 722 | 119 | |
AK314266.1 | - | 3 - 4 | 687 | 119 | |
AY029770.1 | - | 3 - 4 | 654 | 119 | |
AY322551.1 | - | 3 - 4 | 654 | 119 | |
BC000740.2 | - | 3 - 4 | 710 | 119 | |
BT006789.1 | - | 3 - 4 | 654 | 119 | |
CD013884.1 | - | 2 - 3 | 525 | 119 | |
D13305.1 | - | 3 - 4 | 846 | 119 | |
L04473.1 | - | 3 - 4 | 654 | 119 | |
L07746.1 | - | 3 - 4 | 816 | 119 | |
L08112.1 | - | 3 - 4 | 654 | 119 |