The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000779.3 | NP_000770.2 | 4 - 5 | 583 | 85 |
NM_001099772.1 | NP_001093242.1 | 4 - 5 | 583 | 85 | |
NM_001319161.1 | NP_001306090.1 | 3 - 4 | 538 | 85 | |
NM_001319162.1 | NP_001306091.1 | 2 - 3 | 396 | 85 | |
NM_001319163.1 | NP_001306092.1 | 2 - 3 | 396 | 85 | |
NR_135003.1 | 4 - 5 | 583 | 85 | ||
XM_011540832.1 | XP_011539134.1 | 4 - 5 | 586 | 85 | |
XM_011540833.1 | XP_011539135.1 | 3 - 4 | 218 | 85 | |
XM_017000466.1 | XP_016855955.1 | 3 - 4 | 490 | 85 | |
GenBank mRNA | AF491285.1 | - | 4 - 5 | 499 | 85 |
AK225576.1 | - | 4 - 5 | 535 | 85 | |
AK225578.1 | - | 4 - 5 | 535 | 85 | |
AK300921.1 | - | 2 - 3 | 396 | 85 | |
AK300941.1 | - | 2 - 3 | 245 | 85 | |
AK307715.1 | - | 4 - 5 | 657 | 85 | |
AK313170.1 | - | 4 - 5 | 535 | 85 | |
AY064484.1 | - | 4 - 5 | 499 | 85 | |
AY064485.1 | - | 4 - 5 | 499 | 85 | |
AY064486.1 | - | 4 - 5 | 499 | 85 | |
AY151048.1 | - | 4 - 5 | 499 | 85 | |
AY151049.1 | - | 3 - 4 | 454 | 85 | |
BC017758.1 | - | 4 - 5 | 535 | 85 | |
CD013981.1 | - | 4 - 5 | 517 | 85 | |
DC358497.1 | - | 2 - 3 | 348 | 85 | |
J02871.1 | - | 4 - 5 | 511 | 85 | |
X16699.1 | - | 4 - 5 | 499 | 85 |