The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243766.1 | NP_001230695.1 | 9 - 10 | 1532 | 58 |
NM_001290129.1 | NP_001277058.1 | 8 - 9 | 953 | 58 | |
NM_001290130.1 | NP_001277059.1 | 8 - 9 | 919 | 58 | |
NM_017739.3 | NP_060209.3 | 9 - 10 | 1045 | 58 | |
XM_005271010.1 | XP_005271067.1 | 9 - 10 | 1040 | 58 | |
XM_006710755.1 | XP_006710818.1 | 9 - 10 | 1005 | 58 | |
XM_006710756.1 | XP_006710819.1 | 9 - 10 | 1040 | 58 | |
XM_011541760.2 | XP_011540062.1 | 8 - 9 | 897 | 58 | |
XM_017001690.1 | XP_016857179.1 | 9 - 10 | 1005 | 58 | |
GenBank mRNA | AB057356.1 | - | 9 - 10 | 975 | 58 |
AF250859.2 | - | 6 - 7 | 634 | 58 | |
AK000284.1 | - | 9 - 10 | 1022 | 58 | |
AK022727.1 | - | 4 - 5 | 391 | 58 | |
AK056186.1 | - | 9 - 10 | 1532 | 58 | |
AK225033.1 | - | 9 - 10 | 1023 | 58 | |
AK301923.1 | - | 8 - 9 | 919 | 58 | |
AK302379.1 | - | 8 - 9 | 929 | 58 | |
AK316399.1 | - | 8 - 9 | 926 | 58 | |
AY358592.1 | - | 9 - 10 | 1010 | 58 | |
BC001471.2 | - | 9 - 10 | 1039 | 58 | |
CR749700.1 | - | 8 - 9 | 1400 | 58 |