The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193360.1 | NP_001180289.1 | 6 - 7 | 1299 | 95 |
NM_001193361.1 | NP_001180290.1 | 5 - 6 | 1215 | 95 | |
NM_001193362.1 | NP_001180291.1 | 6 - 7 | 1458 | 95 | |
NM_001193363.1 | NP_001180292.1 | 5 - 6 | 1374 | 95 | |
NM_018199.3 | NP_060669.1 | 4 - 5 | 835 | 95 | |
NR_034164.1 | 7 - 8 | 1389 | 95 | ||
NR_034165.1 | 7 - 8 | 1548 | 95 | ||
XM_005267817.3 | XP_005267874.1 | 4 - 5 | 765 | 95 | |
XM_011536908.2 | XP_011535210.1 | 4 - 5 | 853 | 95 | |
XM_017021421.1 | XP_016876910.1 | 5 - 6 | 898 | 95 | |
GenBank mRNA | AK001600.1 | - | 4 - 5 | 763 | 95 |
AK056025.1 | - | 7 - 8 | 1335 | 95 | |
AK292563.1 | - | 6 - 7 | 1458 | 95 | |
AK295601.1 | - | 5 - 6 | 1334 | 95 | |
AL834434.1 | - | 4 - 5 | 757 | 95 | |
BC001962.1 | - | 4 - 5 | 895 | 95 | |
BX647767.1 | - | 5 - 6 | 1165 | 95 | |
CR936728.1 | - | 6 - 7 | 1174 | 95 | |
CR936729.1 | - | 5 - 6 | 1182 | 95 |