The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015557.2 | NP_056372.1 | 36 - 37 | 5357 | 82 |
GenBank mRNA | AB007913.1 | - | 18 - 19 | 2318 | 82 |
AF425231.1 | - | 36 - 37 | 5346 | 82 | |
AK126012.1 | - | 16 - 17 | 2149 | 82 | |
AK127046.1 | - | 28 - 29 | 3964 | 82 | |
AL110281.1 | - | 26 - 27 | 3500 | 82 | |
AL117491.1 | - | 26 - 27 | 3550 | 82 | |
AL137693.1 | - | 26 - 27 | 3478 | 82 |