Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTF1
Entrez Gene ID
4520
Gene Name
metal regulatory transcription factor 1
Gene Aliases
MTF-1, ZRF
Chromosome Location
Chr.1: 37809567 - 37859624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005955.2 NP_005946.2 5 - 6 995 146
XM_011541491.2 XP_011539793.1 5 - 6 1159 146
XM_011541493.2 XP_011539795.1 5 - 6 999 146
XM_011541494.1 XP_011539796.1 6 - 7 1018 146
GenBank mRNA AK225923.1 - 5 - 6 967 146
AK309584.1 - 6 - 7 1018 146
AK314233.1 - 5 - 6 967 146
BC014454.1 - 5 - 6 970 146
X78710.1 - 5 - 6 937 146

View Full Product Details