The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005955.2 | NP_005946.2 | 7 - 8 | 1211 | 136 |
XM_011541491.2 | XP_011539793.1 | 7 - 8 | 1375 | 136 | |
XM_011541493.2 | XP_011539795.1 | 7 - 8 | 1215 | 136 | |
XM_011541494.1 | XP_011539796.1 | 8 - 9 | 1234 | 136 | |
GenBank mRNA | AK309584.1 | - | 8 - 9 | 1234 | 136 |
BC014454.1 | - | 7 - 8 | 1186 | 136 | |
X78710.1 | - | 7 - 8 | 1153 | 136 |