Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTF1
Entrez Gene ID
4520
Gene Name
metal regulatory transcription factor 1
Gene Aliases
MTF-1, ZRF
Chromosome Location
Chr.1: 37809567 - 37859624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005955.2 NP_005946.2 9 - 10 1912 91
XM_011541491.2 XP_011539793.1 9 - 10 2076 91
XM_011541494.1 XP_011539796.1 10 - 11 1935 91
GenBank mRNA AK225923.1 - 9 - 10 1884 91
AK298751.1 - 3 - 4 551 91
AK314233.1 - 9 - 10 1884 91
BC014454.1 - 9 - 10 1887 91
X78710.1 - 9 - 10 1854 91

View Full Product Details