The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099406.1 | NP_001092876.1 | 2 - 3 | 465 | 87 |
NM_012319.3 | NP_036451.3 | 3 - 4 | 1263 | 87 | |
XM_011525900.1 | XP_011524202.1 | 3 - 4 | 1696 | 87 | |
XM_011525901.1 | XP_011524203.1 | 4 - 5 | 2380 | 87 | |
GenBank mRNA | AK299102.1 | - | 2 - 3 | 296 | 87 |
AK301539.1 | - | 2 - 3 | 415 | 87 | |
AK304079.1 | - | 3 - 4 | 1187 | 87 | |
BC039498.1 | - | 2 - 3 | 456 | 87 | |
BX336036.2 | - | 2 - 3 | 234 | 87 | |
U41060.2 | - | 4 - 5 | 1092 | 87 |