The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099406.1 | NP_001092876.1 | 4 - 5 | 856 | 132 |
NM_012319.3 | NP_036451.3 | 5 - 6 | 1654 | 132 | |
XM_011525900.1 | XP_011524202.1 | 5 - 6 | 2087 | 132 | |
XM_011525901.1 | XP_011524203.1 | 6 - 7 | 2771 | 132 | |
GenBank mRNA | AK299102.1 | - | 4 - 5 | 687 | 132 |
AK301539.1 | - | 4 - 5 | 806 | 132 | |
AK304079.1 | - | 5 - 6 | 1578 | 132 | |
BC008317.2 | - | 1 - 2 | 223 | 132 | |
BC039498.1 | - | 4 - 5 | 847 | 132 | |
BX336036.2 | - | 4 - 5 | 625 | 132 | |
U41060.2 | - | 6 - 7 | 1483 | 132 |