Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
Chromosome Location
Chr.2: 96122817 - 96143059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002036.3 NP_001002036.3 5 - 6 455 117
XM_011511205.2 XP_011509507.1 4 - 5 1270 117
XM_011511206.2 XP_011509508.1 5 - 6 1493 117
XM_011511207.2 XP_011509509.1 4 - 5 1271 117
XM_011511208.2 XP_011509510.1 4 - 5 1271 117
GenBank mRNA AJ537600.2 - 5 - 6 455 117
BC107126.1 - 5 - 6 454 117
BC107127.1 - 5 - 6 455 117

View Full Product Details