Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MGA
- Entrez Gene ID
-
23269
- Gene Name
- MGA, MAX dimerization protein
- Gene Aliases
-
MAD5, MXD5
- Chromosome Location
-
Chr.15: 41621139 - 41769943 on Build GRCh38
View Full Product Details