Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMS19
Entrez Gene ID
64210
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases
MET18, MMS19L, hMMS19
Chromosome Location
Chr.10: 97458324 - 97498794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289403.1 NP_001276332.1 9 - 10 948 68
NM_001289404.1 NP_001276333.1 9 - 10 1016 68
NM_001289405.1 NP_001276334.1 11 - 12 959 68
NM_022362.4 NP_071757.4 10 - 11 1077 68
XM_005270035.2 XP_005270092.1 9 - 10 834 68
XM_005270041.1 XP_005270098.1 8 - 9 722 68
XM_006717944.3 XP_006718007.2 10 - 11 871 68
XM_006717945.2 XP_006718008.1 8 - 9 1388 68
XM_011540062.1 XP_011538364.1 9 - 10 680 68
XM_011540063.2 XP_011538365.1 8 - 9 1388 68
XM_017016515.1 XP_016872004.1 11 - 12 983 68
XM_017016516.1 XP_016872005.1 11 - 12 983 68
XM_017016517.1 XP_016872006.1 10 - 11 854 68
XM_017016518.1 XP_016872007.1 9 - 10 742 68
XM_017016519.1 XP_016872008.1 11 - 12 983 68
XM_017016520.1 XP_016872009.1 10 - 11 871 68
XM_017016521.1 XP_016872010.1 10 - 11 871 68
XM_017016522.1 XP_016872011.1 10 - 11 975 68
XM_017016523.1 XP_016872012.1 9 - 10 859 68
XM_017016524.1 XP_016872013.1 9 - 10 834 68
XM_017016525.1 XP_016872014.1 9 - 10 740 68
XM_017016526.1 XP_016872015.1 9 - 10 815 68
XM_017016527.1 XP_016872016.1 9 - 10 834 68
XM_017016528.1 XP_016872017.1 8 - 9 1388 68
XM_017016529.1 XP_016872018.1 9 - 10 1456 68
XM_017016530.1 XP_016872019.1 10 - 11 975 68
XM_017016531.1 XP_016872020.1 9 - 10 834 68
XM_017016532.1 XP_016872021.1 8 - 9 611 68
XM_017016533.1 XP_016872022.1 8 - 9 691 68
XM_017016534.1 XP_016872023.1 8 - 9 1388 68
XM_017016535.1 XP_016872024.1 8 - 9 1388 68
GenBank mRNA AF319947.1 - 11 - 12 925 68
AF357881.1 - 10 - 11 876 68
AJ306408.1 - 10 - 11 859 68
AK027710.1 - 9 - 10 711 68
AK056581.1 - 6 - 7 474 68
AK225312.1 - 10 - 11 862 68
AK298995.1 - 10 - 11 915 68
AK304287.1 - 9 - 10 742 68
AK314374.1 - 10 - 11 875 68
BC006575.2 - 2 - 3 171 68
BC080532.1 - 9 - 10 795 68
BC117129.1 - 10 - 11 875 68

View Full Product Details