Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMS19
Entrez Gene ID
64210
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases
MET18, MMS19L, hMMS19
Chromosome Location
Chr.10: 97458324 - 97498794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289403.1 NP_001276332.1 7 - 8 782 95
NM_001289404.1 NP_001276333.1 7 - 8 850 95
NM_001289405.1 NP_001276334.1 9 - 10 793 95
NM_022362.4 NP_071757.4 8 - 9 911 95
XM_005270035.2 XP_005270092.1 7 - 8 668 95
XM_005270041.1 XP_005270098.1 6 - 7 556 95
XM_006717944.3 XP_006718007.2 8 - 9 705 95
XM_006717945.2 XP_006718008.1 6 - 7 1222 95
XM_011540062.1 XP_011538364.1 7 - 8 514 95
XM_011540063.2 XP_011538365.1 6 - 7 1222 95
XM_017016515.1 XP_016872004.1 9 - 10 817 95
XM_017016516.1 XP_016872005.1 9 - 10 817 95
XM_017016517.1 XP_016872006.1 8 - 9 688 95
XM_017016518.1 XP_016872007.1 7 - 8 576 95
XM_017016519.1 XP_016872008.1 9 - 10 817 95
XM_017016520.1 XP_016872009.1 8 - 9 705 95
XM_017016521.1 XP_016872010.1 8 - 9 705 95
XM_017016522.1 XP_016872011.1 8 - 9 809 95
XM_017016523.1 XP_016872012.1 7 - 8 693 95
XM_017016524.1 XP_016872013.1 7 - 8 668 95
XM_017016525.1 XP_016872014.1 7 - 8 574 95
XM_017016526.1 XP_016872015.1 7 - 8 649 95
XM_017016527.1 XP_016872016.1 7 - 8 668 95
XM_017016528.1 XP_016872017.1 6 - 7 1222 95
XM_017016529.1 XP_016872018.1 7 - 8 1290 95
XM_017016530.1 XP_016872019.1 8 - 9 809 95
XM_017016531.1 XP_016872020.1 7 - 8 668 95
XM_017016532.1 XP_016872021.1 6 - 7 445 95
XM_017016533.1 XP_016872022.1 6 - 7 525 95
XM_017016534.1 XP_016872023.1 6 - 7 1222 95
XM_017016535.1 XP_016872024.1 6 - 7 1222 95
GenBank mRNA AF319947.1 - 9 - 10 759 95
AF357881.1 - 8 - 9 710 95
AJ306408.1 - 8 - 9 693 95
AK027710.1 - 7 - 8 545 95
AK056581.1 - 4 - 5 308 95
AK298995.1 - 8 - 9 749 95
AK304287.1 - 7 - 8 576 95
AK314374.1 - 8 - 9 709 95
BC080532.1 - 7 - 8 629 95
BC117129.1 - 8 - 9 709 95

View Full Product Details