The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098536.1 | NP_001092006.1 | 10 - 11 | 1274 | 126 |
NM_003481.2 | NP_003472.2 | 10 - 11 | 1268 | 126 | |
GenBank mRNA | AK308021.1 | - | 10 - 11 | 1258 | 126 |
BC004889.2 | - | 10 - 11 | 1247 | 126 | |
BC005139.2 | - | 10 - 11 | 1245 | 126 | |
U35116.1 | - | 10 - 11 | 1258 | 126 | |
U47927.1 | - | 10 - 11 | 1267 | 126 | |
X91349.1 | - | 10 - 11 | 1244 | 126 |