The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098536.1 | NP_001092006.1 | 1 - 2 | 170 | 69 |
NM_003481.2 | NP_003472.2 | 1 - 2 | 164 | 69 | |
GenBank mRNA | AK308021.1 | - | 1 - 2 | 154 | 69 |
AU099996.1 | - | 1 - 2 | 171 | 69 | |
BC004889.2 | - | 1 - 2 | 143 | 69 | |
BC005139.2 | - | 1 - 2 | 141 | 69 | |
CN276110.1 | - | 1 - 2 | 164 | 69 | |
U35116.1 | - | 1 - 2 | 154 | 69 | |
U47927.1 | - | 1 - 2 | 163 | 69 |