Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNT1
Entrez Gene ID
7138
Gene Name
troponin T1, slow skeletal type
Gene Aliases
ANM, NEM5, STNT, TNT, TNTS
Chromosome Location
Chr.19: 55132698 - 55149354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126132.2 NP_001119604.1 7 - 8 397 79
NM_001126133.2 NP_001119605.1 6 - 7 364 79
NM_001291774.1 NP_001278703.1 7 - 8 459 79
NM_003283.5 NP_003274.3 7 - 8 397 79
XM_011527246.2 XP_011525548.1 6 - 7 274 79
XM_017027186.1 XP_016882675.1 8 - 9 470 79
XM_017027187.1 XP_016882676.1 7 - 8 458 79
GenBank mRNA AK291352.1 - 7 - 8 249 79
AK308199.1 - 6 - 7 216 79
AY762903.1 - 6 - 7 192 79
AY762904.1 - 5 - 6 159 79
BC007074.1 - 6 - 7 237 79
BC010963.1 - 7 - 8 250 79
BC022086.2 - 7 - 8 298 79
BC034143.1 - 7 - 8 249 79
BT019630.1 - 5 - 6 159 79
DC385627.1 - 7 - 8 397 79
DC385695.1 - 7 - 8 282 79
M19308.1 - 6 - 7 217 79
M19309.1 - 7 - 8 249 79
S51938.1 - 5 - 6 159 79
S69208.1 - 7 - 8 233 79
S69209.1 - 6 - 7 200 79

View Full Product Details