Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP5
Entrez Gene ID
126206
Gene Name
NLR family pyrin domain containing 5
Gene Aliases
CLR19.8, MATER, NALP5, PAN11, PYPAF8
Chromosome Location
Chr.19: 55999726 - 56061810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153447.4 NP_703148.4 8 - 9 2447 92
GenBank mRNA AY054986.1 - 8 - 9 2447 92
AY154460.1 - 8 - 9 2447 92

View Full Product Details