The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142503.2 | NP_001135975.1 | 9 - 10 | 2459 | 63 |
NM_001142504.2 | NP_001135976.1 | 8 - 9 | 2353 | 63 | |
NM_014725.4 | NP_055540.2 | 8 - 9 | 2377 | 63 | |
XM_005262314.4 | XP_005262371.1 | 9 - 10 | 2445 | 63 | |
XM_005262315.3 | XP_005262372.1 | 8 - 9 | 2365 | 63 | |
XM_011531069.2 | XP_011529371.1 | 8 - 9 | 2233 | 63 | |
XM_011531070.2 | XP_011529372.1 | 8 - 9 | 2246 | 63 | |
GenBank mRNA | AK291747.1 | - | 8 - 9 | 2353 | 63 |
BC035587.1 | - | 8 - 9 | 2388 | 63 | |
CR749411.1 | - | 9 - 10 | 2360 | 63 | |
D80011.1 | - | 8 - 9 | 2342 | 63 |