Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SREBF1
Entrez Gene ID
6720
Gene Name
sterol regulatory element binding transcription factor 1
Gene Aliases
SREBP-1c, SREBP1, SREBP1a, bHLHd1
Chromosome Location
Chr.17: 17811349 - 17837017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005291.2 NP_001005291.1 16 - 17 3022 123
NM_001321096.2 NP_001308025.1 15 - 16 2794 123
NM_004176.4 NP_004167.3 15 - 16 2932 123
XM_005256772.4 XP_005256829.1 15 - 16 2932 123
XM_017024970.1 XP_016880459.1 13 - 14 2363 123
XM_017024971.1 XP_016880460.1 11 - 12 1714 123
GenBank mRNA AB209609.1 - 14 - 15 3398 123
AK091131.1 - 15 - 16 2761 123
AK095325.1 - 10 - 11 1771 123
AK128320.1 - 13 - 14 2441 123
AK293795.1 - 15 - 16 2632 123
AK294800.1 - 16 - 17 2610 123
AK297113.1 - 13 - 14 2346 123
BC023621.2 - 9 - 10 1356 123
BC026962.1 - 9 - 10 1548 123
BC057388.1 - 15 - 16 2923 123
BC063281.1 - 16 - 17 2997 123
U00968.1 - 15 - 16 2904 123

View Full Product Details